Oakton® pH Buffer Capsules
Fresh pH calibration solution anytime and anywhere
Buffer salts inside capsules require 100 mL of deionized or distilled water to produce a clear pH calibration solution accurate to ±0.02 pH (at 25°C) with traceability to NIST standards. Capsules are easier to store and transport than prepared aqueous buffer solutions. Capsules are color-coded for easy identification if mixed.
Store buffer solution in an airtight container at room temperature for three weeks. Unused capsules have a five-year shelf life. Each vial contains 10 capsules.
$34.92 - $37.13CAD / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Buffer Capsules, pH 4; 10/Pk
$37.13CAD / Pkg of 10
COMPARE
Buffer Capsules, pH 7; 10/Pk
$37.13CAD / Pkg of 10
COMPARE
Buffer Capsules, pH 10; 10/Pk
$34.92CAD / Pkg of 10