This product is now discontinued, View Related Products
Nor-Lake Scientific General Purpose Freezers
Full-sized, single-compartment units cover all your storage needs
  • Operating temperature of -10°C to -20°C
These general purpose freezers feature CFC-free insulation and coolant and manual temperature control. Easily identify samples within your freezer with the interior LED light.
$3,816.08 - $4,744.32CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Freezer Capacity (cu ft) Description
Availability Pricing
Nor-Lake LF161WWW/0M Upright Freezer, Manual Defrost, 17.1 CuFt; 115 VAC RK-44173-01 Mfr # LF161WWW//0M
COMPARE
17.1 Upright Freezer, Manual Defrost, 17.1 CuFt; 115 VAC
$3,816.08CAD / Each
Nor-Lake LF201WWW/0 Upright Freezer, Auto Defrost, 20 CtFt; 115 VAC RK-44173-03 Mfr # LF201WWW/0
COMPARE
20.3 Upright Freezer, Auto Defrost, 20 CtFt; 115 VAC
$4,744.32CAD / Each
Nor-Lake LF201WWW/0M Upright Freezer, Manual Defrost, 20.6 CuFt; 115 VAC RK-44173-05 Mfr # LF201WWW/0M
COMPARE
20.6 Upright Freezer, Manual Defrost, 20.6 CuFt; 115 VAC
$4,272.83CAD / Each