This product is now discontinued, View Related Products
Nor-Lake Premier Laboratory Refrigerators
Maximum capacity for storage of your valuable samples
  • Energy Star models conserve energy and save money
  • Standard lock keeps samples secure
  • Top-mounted compressor maximizes interior storage space
Heavy duty refrigeration system is top mounted to maximize your storage space. Forced air flow is ideal for temperature uniformity throughout the unit. LED microprocessor control features adjustable temperature from 2 to 10°C and includes both an audible and visual alarm with remote contacts, to ensure your samples remain at your desired temperature. Constructed of white epoxy coated galvinized steel, the high density polyurethane foam insulation helps maintain the set temperature.
Solid-door units are Energy Star rated for maximum energy conservation. The glass-door units feature full size, double pane glass with anti-condensate heating wires to maintain full view of samples at all time. All models feature an interior light which is switch activated for easier viewing of your samples.
$4,888.71 - $15,175.92CAD / Each
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
Compareitem specifications
Number of Doors Power (VAC) Refrigerator Capacity (cu ft)
Availability Pricing
Nor-Lake Premier Refrigerator, Sliding Glass Door, 48.75 cu ft, 115V, 60 Hz RK-44173-08 Mfr # NSPR502WWG/0
COMPARE
2 115 48.75
$12,892.16CAD / Each
Nor-Lake Premier Refrigerator, Solid Door, 24 cu ft, 115V, 60 Hz RK-44173-76 Mfr # NSPR241WWW/0
COMPARE
1 115 24
$7,131.21CAD / Each
Nor-Lake Premier Refrigerator, Solid Door, 24 cu ft, 230V, 50 Hz RK-44173-78 Mfr # NSPR241WWW/5
COMPARE
1 230 24
$7,193.09CAD / Each
COMPARE
2 230 48.75
$12,389.74CAD / Each
Nor-Lake Premier Refrigerator, Solid Door, 52 cu ft, 115V, 60 Hz RK-44173-80 Mfr # NSPR522WWW/0
COMPARE
2 115 52
$10,337.30CAD / Each
Nor-Lake Premier Refrigerator, Solid Door, 33 cu ft, 115V, 60 Hz RK-44173-81 Mfr # NSPR331WWW/0
COMPARE
1 115 33
$8,684.16CAD / Each
Nor-Lake Premier Refrigerator, Solid Door, 52 cu ft, 230V, 50 Hz RK-44173-82 Mfr # NSPR522WWW/5
COMPARE
2 230 52
$10,132.50CAD / Each
Nor-Lake Premier Refrigerator, Solid Door, 33 cu ft, 230V, 50 Hz RK-44173-83 Mfr # NSPR331WWW/5
COMPARE
1 230 33
$8,684.16CAD / Each
Nor-Lake Premier Refrigerator, LED Display with Alarm, 80 cu ft with Casters, 115V/208-230V/60Hz RK-44173-84 Mfr # NSPR803WWW/8
COMPARE
2 solid 230
$10,774.90CAD / Each
Nor-Lake Premier Refrigerator, Solid Door, 80 cu ft, 115V, 60 Hz RK-44173-85 Mfr # NSPR803WWW/0
COMPARE
3 115 80
$13,393.12CAD / Each
Nor-Lake Premier Refrigerator, Solid Door, 80 cu ft, 230V, 50 Hz RK-44173-86 Mfr # NSPR803WWW/5
COMPARE
3 230 80
$13,393.12CAD / Each
Nor-Lake Premier Refrigerator, Glass Door, 33 cu ft, 115V, 60 Hz RK-44173-87 Mfr # NSPR331WWG/0
COMPARE
1 115 33
$9,712.59CAD / Each
Nor-Lake Premier Refrigerator, Glass Door, 24 cu ft, 115V, 60 Hz RK-44173-88 Mfr # NSPR241WWG/0
COMPARE
1 115 24
$7,929.78CAD / Each
Nor-Lake Premier Refrigerator, Glass Door, 33 cu ft, 230V, 50 Hz RK-44173-89 Mfr # NSPR331WWG/5
COMPARE
1 230 33
$9,712.59CAD / Each
Nor-Lake Premier Refrigerator, Glass Door, 24 cu ft, 230V, 50 Hz RK-44173-90 Mfr # NSPR241WWG/5
COMPARE
1 230 24
$7,145.94CAD / Each
Nor-Lake Premier Refrigerator, Glass Door, 80 cu ft, 115V, 60 Hz RK-44173-91 Mfr # NSPR803WWG/0
COMPARE
3 115 80
$15,175.92CAD / Each
Nor-Lake Premier Refrigerator, Glass Door, 52 cu ft, 115V, 60 Hz RK-44173-92 Mfr # NSPR522WWG/0
COMPARE
2 115 52
$11,420.25CAD / Each
Nor-Lake Premier Refrigerator, Glass Door, 52 cu ft, 230V, 50 Hz RK-44173-94 Mfr # NSPR522WWG/5
COMPARE
2 230 52
$11,420.25CAD / Each
Nor-Lake Premier Refrigerator, LED Display with Alarm, 3 Glass Doors, 80 cu ft with Casters, 115V/208-230V/60Hz RK-44173-96 Mfr # NSPR803WWG/8
COMPARE
2 115
$12,215.88CAD / Each
Nor-Lake Premier Refrigerator, Glass Door, 80 cu ft, 230V, 50 Hz RK-44173-98 Mfr # NSPR803WWG/5
COMPARE
3 230 80
$15,175.92CAD / Each
Showing 1 - 20 of 21