This product is now discontinued, View Related Products
Nor-Lake Flammable Storage Units
Sparkfree storage for your flammable products
Heavy duty refrigeration system is top mounted to maximize your storage space. Choose refrigerator for storage from 2 to 10-degrees Celsius, or freezer for storage from -25 to -10-degrees Celsius. Constructed of white epoxy coated galvinized steel, the high density polyurethane foam insulation helps maintain the set temperature. Interior liner and door panel are smooth aluminum, making for easy clean-up.
Solid-door units are Energy Star rated for maximum energy conservation. The cold wall evaporator system eliminates build up of condensation. Manual defrost minimizes the flucuation of temperatures within the unit to keep your samples at your set temperature. Four casters are standard for easy movement. Door lock protects your samples.
$7,747.08 - $8,010.82CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Freezer Capacity (cu ft) Refrigerator Capacity (cu ft) Description
Availability Pricing
Nor-Lake NSFF241WMW/0M Freezer, Flammable Storage 24 cu ft with Casters, 115V/60Hz RK-44173-48 Mfr # NSFF241WMW/0M
COMPARE
24 Freezer, Flammable Storage 24 cu ft with Casters, 115V/60Hz
$8,010.82CAD / Each
Nor-Lake NSFR241WMW/0M Refrigerator, Flammable Storage 24 cu ft with Casters, 115V/60Hz RK-44173-50 Mfr # NSFR241WMW/0M
COMPARE
55 24 Refrigerator, Flammable Storage 24 cu ft with Casters, 115V/60Hz
$7,747.08CAD / Each