This product is now discontinued, View Related Products
Nonconductive Instrument Carts 2770
Transport instruments without fear of static discharge
  • Casters cushion instruments during transport
  • Ribbed-vinyl shelf surfaces eliminates static electricity
  • Capacity is 600 lb (272 kg)
Nonconductive Instrument Cart features ribbed-vinyl shelf surfaces to eliminate static electricity and cushion instruments during transport. Bumper protects walls. Carts 47352-00 and -10 have shock- absorbing, 8" dia pneumatic casters (two swivel). Cart 47352-20 has 5" dia polyurethane casters (all swivel) with brakes. Drawer on 47352-10 locks. Top shelf measures 36"L x 22"W; bottom shelf measures 30"L x 20"W. Overall height is 34". Capacity is 600 lb (272 kg).
$714.30 - $1,272.64CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (lbs) Number of Shelves Shelf Height (in)
Availability Pricing
Brennan MIC2436 Nonconductive Instrument Cart, 8" Casters, 36x20"/30x20", 600 lbs Cap RK-47352-00 Mfr # MIC2436
COMPARE
600 2
$839.46CAD / Each
Brennan MIC2436-5-BRK Nonconductive instrument cart with brakes RK-47352-20 Mfr # MIC2436-5-BRK
COMPARE
600 2
$714.30CAD / Each
Brennan MIC-8BK-PSDR Cart with outlet strip and drawer, two shelf, 8" x 3" casters RK-47355-24 Mfr # MIC-8BK-PSDR
COMPARE
600 2
$1,272.64CAD / Each
Brennan MIC-5BK-PSDR Cart with outlet strip and drawer, two shelf, 5" x 1 1/4" casters RK-47355-26 Mfr # MIC-5BK-PSDR
COMPARE
600 2
$1,050.90CAD / Each