Sherwood Scientific Fluid Bed Dryer Non-Sealed Tub Assemblies
Quickly and evenly dry particles as small as 5 μm
  • Faster and gentler than conventional drying cabinets—ideal for food products, polymers, soils, chemicals, pharmaceuticals, minerals, and waste materials
  • Variety of accessories provide easy customization for your drying applications
Non-Sealed Tub Assemblies are available in glass or stainless steel and require a filter bag, sold separately below.
Model 02658-02 includes a port to add extra sample and a port for a temperature/humidity probe (sold separately).
Order fluid bed dryer, filter bags, and optional accessories separately for a complete fluid bed dryer system.
$1,126.41 - $3,889.75CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Materials of Construction Description
Availability Pricing
Sherwood Scientific 50035010 Tub assembly, non-sealed, 5 L, with port for temperature/humidity probe RK-02658-02 Mfr # 50035010
COMPARE
Glass Tub assembly, non-sealed, 5 L, with port for temperature/humidity probe
$3,889.75CAD / Each
Sherwood Scientific 50035006 Fluid Bed Dryer Tub and Assembly, Non-Sealed, Glass, 2-Liter RK-02658-52 Mfr # 50035006
COMPARE
Glass Fluid Bed Dryer Tub and Assembly, Non-Sealed, Glass, 2-Liter
$2,880.48CAD / Each
Sherwood Scientific 50035009 Fluid Bed Dryer Tub and Assembly, Non-Sealed, Glass, 5-Liter RK-02658-56 Mfr # 50035009
COMPARE
Glass Fluid Bed Dryer Tub and Assembly, Non-Sealed, Glass, 5-Liter
$3,646.64CAD / Each
Sherwood Scientific 50035013 Tub assembly, non-sealed, 4 x 250 mL, glass RK-02658-62 Mfr # 50035013
COMPARE
Glass Tub assembly, non-sealed, 4 x 250 mL, glass
$1,126.41CAD / Each