This product is now discontinued, View Related Products
Non-Electric Steam Sterilizers
Provide complete, efficient steam sterilization at low cost
Non-electric steam sterilizers provide effective sterilization at a low cost! With an efficient heat source and a small amount of water, these sterilizers produce enough steam at 15 psi to destroy any type of bacteria or micro-organisms. These sterilizers are low maintenance and require no accessories. Safety clamp locks prevent removal of the cover before the pressure is released.
$811.84 - $1,554.43CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Chamber Capacity (Qt)
Availability Pricing
COMPARE
9.5
$811.84CAD / Each
COMPARE
17.6
$870.77CAD / Each
COMPARE
27.9
$1,554.43CAD / Each