Myron L pH UltraPen Testers
Waterproof, pocket-sized meters for water analysis testing
  • Up to 3 point calibration with 4, 7 and 10 buffer
  • Automatic temperature compensation
  • PTBT2 model with Bluetooh for data transfer
These waterproof pH and ORP testers offer anodized aircraft aluminum with protective coating and fully encapsulated electronics for durability. The LCD provides measurement reading with temperature available in both a live and hold measurement mode. Calibration features three point automatic buffer recognition for pH 4, 7, and 10 buffers.
$428.02 - $749.22CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Myron L UltraPen PT2 UltraPen 2 pH and Temperature Pen with Bluetooth RK-05754-35 Mfr # PTBT2
COMPARE
UltraPen 2 pH and Temperature Pen with Bluetooth
$749.22CAD / Each
COMPARE
UltraPen pH and Temperature Pen
$428.02CAD / Each
COMPARE
UltraPen ORP and Temperature Pen
$459.70CAD / Each