MultiMagStir Genie Progammable Multi-position Stirrers
Program parameters for up to nine distinct levels and store profiles in memory
  • Low-pofile design conserves precious laboratory bench space
  • Three power levels handle various viscosities
This four-position stirrer can be used at both high and low speed ranges with up to four 1-L beakers or three 1-L spinner flasks. Microprocessor-controlled electronics ensure precise, reproducible results over time without heat buildup or internal friction.
Versatile stirrer features four programming modes with digital display of setup and readings. Operate unit in manual mode for basic stir, reverse, and timed stirring functions. Setup mode is used to program and edit default parameters for manual operation. Automatic mode activates one of four user-defined profiles previously stored in memory. Profile mode lets you create, edit, and store up to four customized profiles; each profile can consist of up to nine independent stirring steps.
$1,723.87CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Scientific Industries Genie SI-3000 MultiMagStir , Programmable 4-Position Stirrer; 120V RK-86575-00 Mfr # SI-3000
COMPARE
120
$1,723.87CAD / Each