This product is now discontinued, View Related Products
Motic Moticam Pro Digital Microscope Cameras
Capture high-quality images with this scientific-grade CCD camera
Internal 4-frame buffer provides faster and more consistent frame rates. On-board image processing allows you to apply white balance or color corrections before the image is sent to your computer. Capture live, still, time-lapse, and video images with the included Motic Images Advanced 3.2 software. Software features include measurement, editing, amalgamation, manual/single point segmentation, single image assembly from multiple images, and multi-language display. USB 2.0 output provides easy communication with your PC or laptop and quick plug-and-play setup.
$3,765.25 - $6,146.25CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Image Sensor Power Resolution
Availability Pricing
Motic Instruments 205A Digital Microscope Camera, 1/2" CCD RK-03908-71 Mfr # 205A
COMPARE
1/2\" CCD USB 1360 x 1024
$3,765.25CAD / Each
Motic Instruments 252A am Pro Digital Microscope Camera, 1/1.8" CCD RK-03908-72 Mfr # 252A
COMPARE
1/1.8\" CCD USB 2080 x 1542
$4,069.50CAD / Each
Motic Instruments 282A Digital Microscope Camera, 2/3" CCD RK-03908-73 Mfr # 282A
COMPARE
2/3\" CCD USB 2580 x 1944
$5,685.08CAD / Each
Motic Instruments 205B Digital Microscope Camera, 1/2" CCD, Peltier cooled RK-03908-74 Mfr # 205B
COMPARE
1/2\" CCD USB (camera) and VAC (cooling) 1360 x 1024
$4,523.31CAD / Each
Motic Instruments 252B Digital Microscope Camera, 1/1.8" CCD, Peltier cooled RK-03908-75 Mfr # 252B
COMPARE
1/1.8\" CCD USB (camera) and VAC (cooling) 2080 x 1542
$4,685.38CAD / Each
Motic Instruments 282B Digital Microscope Camera, 2/3" CCD, Peltier cooled RK-03908-76 Mfr # 282B
COMPARE
2/3\" CCD USB (camera) and VAC (cooling) 2580 x 1944
$6,146.25CAD / Each