This product is now discontinued, View Related Products
Modular Homogenizer Shafts with Generator Assemblies
Choose the Shaft and Generator Assembly that fits your sample size and viscosity
  • Quick-connect shafts and generator assemblies are interchangeable
  • Made from 304 SS and easy to disassemble for cleaning or replacement
Generator assemblies are made up of the rotor and stator. Slots on the rotor and stator have beveled edges like a scissor giving you better shearing action. Unique slanted slots on the rotor constantly draw media through and allow it to build up speed. This eliminates the stop/start effect that slows the media — giving you quicker, more efficient processing.
$726.38 - $1,278.90CAD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Dispersion Type Max Sample Size (mL) Min Sample Size (mL)
Availability Pricing
COMPARE
Fine 100 1
$735.22CAD / Each
COMPARE
Fine 500 5
$777.95CAD / Each
COMPARE
Fine 2000 10
$836.89CAD / Each
COMPARE
Fine 5000 100
$1,278.90CAD / Each
COMPARE
Normal 100 1
$735.22CAD / Each
COMPARE
Normal 500 5
$777.95CAD / Each
COMPARE
Normal 2000 10
$836.89CAD / Each
COMPARE
Normal 5000 30
$1,032.85CAD / Each
COMPARE
Coarse 100 1
$735.22CAD / Each
COMPARE
Coarse 500 5
$786.79CAD / Each
COMPARE
Coarse 2000 10
$836.89CAD / Each
COMPARE
Coarse 5000 30
$1,032.85CAD / Each
JetCat USA 60426 Shaft and Generator Assembly, 1 to 100 mL, Fine viscosity RK-36904-52 Mfr # 60426
COMPARE
Fine 100 1
$726.38CAD / Each
COMPARE
Coarse 2000 30
$1,260.12CAD / Each
Showing 1- 14 of 14