MLA Macro Fixed-Volume Pipettors
Ideal for tasks requiring high accuracy and large volumes
  • Convenient, color-coded, fixed-volume pipets
  • High accuracy and precision
  • Simple single-stroke procedure
Reconstitute your reagents, controls, or dilutions using these precise MLA Macro Pipettors. Calibrated "to deliver" instruments—they have a fixed stroke and consistently deliver the stated or calibrated volume when the plunger is fully depressed. Each pipet can be adjusted to deliver above or below its stated volume within ±10%. Calibration is accomplished readily by inserting a small key into the plunger and turning the key.
These pipettes are color-coded for easy identification and come with the stated volume engraved onto the piston assembly. Constructed of aluminum alloy and stainless steel, these pipettors withstand years of use and provide exceptional performance for a wide range of fluids — including dense or viscous samples.
$422.86 - $518.63CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL)
Availability Pricing
VistaLab MLA Macro 1065 Fixed Volume Pipette, 1.0 mL, Grey; Each RK-06199-15 Mfr # 1065
COMPARE
1
$436.12CAD / Each
VistaLab MLA Macro 1066 Fixed Volume Pipette, 2.0 mL, Red; Each RK-06199-17 Mfr # 1066
COMPARE
2
$422.86CAD / Each
VistaLab MLA Macro 1067 Fixed Volume Pipette, 3.0 mL, Green; Each RK-06199-19 Mfr # 1067
COMPARE
3
$436.12CAD / Each
VistaLab MLA Macro 1068 Fixed Volume Pipette, 4.0 mL, White; Each RK-06199-21 Mfr # 1068
COMPARE
4
$422.86CAD / Each
VistaLab MLA Macro 1069 Fixed Volume Pipette, 5.0 mL, Yellow; Each RK-06199-23 Mfr # 1069
COMPARE
5
$436.12CAD / Each
VistaLab MLA Macro 1070 Fixed Volume Pipette, 10.0 mL, Red; Each RK-06199-25 Mfr # 1070
COMPARE
10
$503.90CAD / Each
VistaLab MLA Macro 1076 Fixed Volume Pipette, 6.0 mL, Red; Each RK-06199-27 Mfr # 1076
COMPARE
6
$503.90CAD / Each
VistaLab MLA Macro 1077 Fixed Volume Pipette, 7.0 mL, Red; Each RK-06199-29 Mfr # 1077
COMPARE
7
$503.90CAD / Each
VistaLab MLA Macro 1078 Fixed Volume Pipette, 8.0 mL, Red; Each RK-06199-31 Mfr # 1078
COMPARE
8
$503.90CAD / Each
VistaLab MLA Macro 1079 Fixed Volume Pipette, 9.0 mL, Red; Each RK-06199-33 Mfr # 1079
COMPARE
9
$518.63CAD / Each
Showing 1- 10 of 10