Microslides and Chamber for BTX Electroporation Systems
Microslides are used primarily with electro cell fusion system. The microslide fits on the microscope allowing observation of the alignment and fusion process. Main applications include embryo manipulation and cell or protoplast fusion. Use with coaxial banana plug connection cable 02894-68 and square post cables 02894-54.
Meander Fusion Chamber 02894-50 is a microslide with a novel design specifically used for electro cell fusion. This chamber is constructed of a conductive metal alloy, which has been deposited in a finger-like projection array on a glass microscope slide. It generates an inhomogenous field and is used for cell fusion of microorganisms, plant, fungi, yeast and mammalian cells.
$4,442.27CAD / Pkg of 10
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
High Voltage Mode - Capacitance High Voltage Mode - Pulse Length High Voltage Mode - Voltage
Availability Pricing
BTX 45-0104 Electroporation Microslide/Microplate, 1 mm Gap, 40 uL; 10/Pk RK-02894-44 Mfr # 45-0104
COMPARE
$4,442.27CAD / Pkg of 10