This product is now discontinued, View Related Products
MicroGuard CE Cleanroom Coveralls
Provides superior worker comfort that is also strong and durable
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
Constructed of a soft, non-woven polypropylene combined with a microporous film, coveralls provide a superior liquid barrier, particulate barrier, and strong tear resistance. The microporous film provides enhanced breathability while the non-woven fabric presents a soft interior for increased worker comfort. Bound seams and tunnelized elastic ensure extremely low linting. Clean processed and individually packaged.
Compliance: ISO Class 5
$291.73 - $456.75CAD / Case of 25
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Size Description
Availability Pricing
International Enviroguard Cleanroom Coveralls, elastic wrist and ankle, medium RK-33683-11 Mfr # CE8013-CI-M
COMPARE
Medium Cleanroom Coveralls, elastic wrist and ankle, medium
$291.73CAD / Case of 25
International Enviroguard Cleanroom Coveralls, elastic wrist and ankle, large RK-33683-12 Mfr # CE8013-CI-L
COMPARE
Large Cleanroom Coveralls, elastic wrist and ankle, large
$291.73CAD / Case of 25
International Enviroguard Cleanroom Coveralls, elastic wrist and ankle, extra large RK-33683-13 Mfr # CE8013-CI-XL
COMPARE
X-large Cleanroom Coveralls, elastic wrist and ankle, extra large
$291.73CAD / Case of 25
International Enviroguard Cleanroom Coveralls, Elastic Wrist/Ankle, XXL; 25/Cs RK-33683-14 Mfr # CE8013-CI-2XL
COMPARE
XX-large Cleanroom Coveralls, Elastic Wrist/Ankle, XXL; 25/Cs
$291.73CAD / Case of 25
International Enviroguard Cleanroom Coveralls, Elastic Wrist/Ankle, Hood/Boot, M; 25/Cs RK-33683-21 Mfr # CE8019-CI-M
COMPARE
Medium Cleanroom Coveralls, Elastic Wrist/Ankle, Hood/Boot, M; 25/Cs
$456.75CAD / Case of 25
International Enviroguard Cleanroom Coveralls, Elastic Wrist/Ankle, Hood/Boot, L; 25/Cs RK-33683-22 Mfr # CE8019-CI-L
COMPARE
Large Cleanroom Coveralls, Elastic Wrist/Ankle, Hood/Boot, L; 25/Cs
$456.75CAD / Case of 25
International Enviroguard Cleanroom Coveralls, Elastic Wrist/Ankle, Hood/Boot, XL; 25/Cs RK-33683-23 Mfr # CE8019-CI-XL
COMPARE
X-Large Cleanroom Coveralls, Elastic Wrist/Ankle, Hood/Boot, XL; 25/Cs
$456.75CAD / Case of 25
International Enviroguard Cleanroom Coveralls, Elastic Wrist/Ankle, Hood/Boot, XXL; 25/Cs RK-33683-24 Mfr # CE8019-CI-2XL
COMPARE
XX-Large Cleanroom Coveralls, Elastic Wrist/Ankle, Hood/Boot, XXL; 25/Cs
$456.75CAD / Case of 25
Showing 1- 8 of 8