This product is now discontinued, View Related Products
Micro Green Clean Biodegradable Cleaner
A free-rinsing, industrial strength, biodegradable hard surface cleaner designed for use in a wide range of applications.
Excellent for cleaning metals, ceramics, medical instruments, food-processing equipment, filter membranes, and other surfaces. Typical pH: 9.5.
$249.00 - $2,301.44CAD / Case of 12
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Dilution Volume (Liters)
Availability Pricing
International Products Corp M-1050-12 Micro Green Clean Biodegradable Cleaner, 12 x 1 L RK-18100-08 Mfr # M-1050-12
COMPARE
1:100 to 2:100, pH 9.8 1
$249.00CAD / Case of 12
International Products Corp M-1052 Micro Green Clean Biodegradable Cleaner, 20.5 KG (19 L) RK-18100-09 Mfr # M-1052
COMPARE
1:100 to 2:100, pH 9.8
$334.46CAD / Each
International Products Corp M-1053 Micro Green Clean Biodegradable Cleaner, 215 KG (198 L) RK-18100-12 Mfr # M-1053
COMPARE
1:100 to 2:100, pH 9.8
$2,301.44CAD / Each