This product is now discontinued, View Related Products
Mettler Toledo WeightLink Test Weights
Designed for positive tolerances
  • High grade stainless steel offers uniform consistency and resistance to corrosion
  • Unique identification number on base links to certificate
These high grade stainless steel WeightLink test weights, combined with a data matrix code (DMC) scanner (sold separately), are part of an innovative, fool-proof identification and traceability weight verification system for balance testing. The weights range from 1 to 100 g with a unique identification number imbedded on their base linked to a calibration certificate, and a calibration eData card. They come with a lifetime warranty.
$129.22 - $176.07CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Mass Class Mass Size (g) Description
Availability Pricing
Mettler Toledo WeightLink 1G Test Weight RK-01030-31 Mfr # 30293487
COMPARE
Class 1 1 1G Test Weight
$129.22CAD / Each
Mettler Toledo WeightLink 2G Test Weight RK-01030-32 Mfr # 30293488
COMPARE
Class 1 2 2G Test Weight
$129.22CAD / Each
Mettler Toledo WeightLink 5G Test Weight RK-01030-33 Mfr # 30293489
COMPARE
Class 1 5 5G Test Weight
$129.22CAD / Each
Mettler Toledo WeightLink 10G Test Weight RK-01030-34 Mfr # 30293490
COMPARE
Class 1 10 10G Test Weight
$129.22CAD / Each
Mettler Toledo WeightLink 20G Test Weight RK-01030-35 Mfr # 30293491
COMPARE
Class 1 20 20G Test Weight
$129.22CAD / Each
Mettler Toledo WeightLink 50G Test Weight RK-01030-36 Mfr # 30293492
COMPARE
Class 1 50 50G Test Weight
$153.23CAD / Each
Mettler Toledo WeightLink 100G Test Weight RK-01030-37 Mfr # 30293493
COMPARE
Class 1 100 100G Test Weight
$176.07CAD / Each
Showing 1- 7 of 7