This product is now discontinued, View Related Products
METTLER TOLEDO One Click Digital Melting Point Apparatus
Video recording and playback on the instrument display – never miss a melting point!
  • Color touch screen with One Click™ user interface
  • Process four samples at once for increased efficiency
  • Conforms to European and US Pharmacopeia (USP) and Japanese Industry Standards (JIS K0064)
Ideal for use in chemical synthesis laboratories, pharmaceutical research and more, this melting point apparatus allows manual or automatic determination of melting point. Program up to twelve One Click™ shortcuts for increased efficiency on repetitive measurements. After entering a program, just a push of a button starts the program, freeing you for other important tasks as the program runs. Run up to four samples at once and display a maximum of 30 minutes of grayscale video on the instrument touch screen to investigate clear points and decomposition temperature. Video magnification of 6x allows easy viewing of smaller samples.
Ergonomic features include small instrument footprint, touch screen controls, and rotatable protection lid. Check your sample for conformity using the substance database incorporated within the unit. Capture statistics such as mean value, min/max, and standard deviation. Print your data using the optional printer for GLP-compliant paper recording.
$1,057.89 - $13,553.71CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Heat-Up Time Max Temperature (° C) Power (VAC)
Availability Pricing
Mettler Toledo MP50 One-Click Digital Melting Point Apparatus; 100-240 VAC RK-03014-20 Mfr # 51142501
COMPARE
0.1° to 20°C per minute (variable) 300 100 to 240
$13,553.71CAD / Each
Mettler Toledo USB-P25 Optional Compact Printer with USB Interface RK-03014-63 Mfr # USB-P25
COMPARE
$1,057.89CAD / Each
Mettler Toledo MP70 One Click Digital Melting Point Apparatus RK-33333-33 Mfr # 51142502
COMPARE
0.1° to 20°C per minute (variable) 350 100 to 240