Mettler Toledo NewClassic MS-TS Analytical Balances
Fast settling time and consistent results for routine to complicated weighing procedures
  • Extra-large color TFT touchscreen display with intuitive user interface
  • Touchscreen can be operated with cotton, silicon and rubber gloves
  • LevelControl warning the user when the balance is not level and provides onscreen help to correct
  • Advanced MonoBloc weighing technology provides fast and accurate results
  • SmartTrac software displays how much of the entire weighing range has been used
  • MinWeigh function displays the weight value in red until the net sample is above the pre-programmed minimum value
  • Passcodes restrict modification of the balance settings to authorized personnel only
  • ISOLog function maintains a record of all weighing relevant changes to the balance’s settings
  • Fully automatic time- and temperature-controlled internal calibration adjustment
  • Durable high-grade chemical-resistant die-cast aluminum housing
  • Overload Protection guards the weighing cell against excess weight overload
  • Direct connection to a PC or printer via RS232 or USB slot allows convenient transfer of data
  • Remove, clean and replace all of the draft shield panels in just a few simple steps, without tools
$10,549.47 - $13,363.65CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Mettler Toledo MS104TS Newclassic MS-TS Analytical Balance, 120g x 0.1 Mg RK-11335-44 Mfr # MS104TS
COMPARE
Internal 120 0.0001
$10,549.47CAD / Each
Mettler Toledo MS204TS Newclassic MS-TS Analytical Balance, 220g x 0.1 Mg RK-11335-45 Mfr # MS204TS
COMPARE
Internal 220 0.0001
$12,258.60CAD / Each
Mettler Toledo MS304TS Newclassic MS-TS Analytical Balance, 320g x 0.1 Mg RK-11335-46 Mfr # MS304TS
COMPARE
Internal 320 0.0001
$13,363.65CAD / Each