Mettler Toledo NewClassic ML-T Analytical Balances
Designed for mobility and easy operation
  • Extra-large color TFT touch-screen display with intuitive user interface
  • Touch-screen can be operated with cotton, silicone, and rubber gloves
  • AC or battery operable for up to 8 hours—ideal for work in field or whenever a power supply is hard to find
  • Fully automatic calibration adjustment (FACT)
  • LevelControl warns you when the balance is not level and offers onscreen help
  • Advanced MonoBloc weighing technology provides fast and accurate results
  • MinWeigh function displays the weight value in red until the net sample is above the preprogrammed minimum value
  • Pass codes restrict modification of the balance settings to authorized personnel only
  • Overload protection guards the weighing cell against excess weight overload
  • Direct connection to a PC or printer via RS-232 or USB slot allows convenient transfer of data
  • Easy-clean feature saves time and effort with all washable parts being dishwasher-safe
  • Remove, clean and replace all of the draft shield panels in just a few simple steps, without tools
$8,295.19 - $10,880.99CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Mettler Toledo ML54T Newclassic Analytical Balance, 52g x 0.1mg RK-11337-00 Mfr # ML54T
COMPARE
Internal 52 0.0001
$8,295.19CAD / Each
Mettler Toledo ML104T Newclassic Analytical Balance, 120G x 0.1mg RK-11337-01 Mfr # ML104T
COMPARE
Internal 120 0.0001
$8,936.11CAD / Each
Mettler Toledo ML204T Newclassic Analytical Balance, 220G x 0.1mg RK-11337-02 Mfr # ML204T
COMPARE
Internal 220 0.0001
$10,173.76CAD / Each
Mettler Toledo ML304T Newclassic Analytical Balance, 320G x0.1mg RK-11337-03 Mfr # ML304T
COMPARE
Internal 320 0.0001
$10,880.99CAD / Each