This product is now discontinued, View Related Products
Mettler Toledo ExcellencePlus XPE Semi-Micro, and Micro Balances
Deliver the highest accuracy for the smallest minimal weight
  • StatusLight™ displays a green light when the balance is ready, yellow for warnings, and red for errors
  • Fully Automatic Calibration Technology (proFACT) performs internal calibrations automatically and at pre-programmed intervals
  • SmartTrac program facilitates dosing in small increments to a target weight
  • Draft shield opens manually or automatically with the push of a button, and dismantles easily for cleanup
  • SmartGrid weighing platform reduces effect of turbulence, shortening stabilization time
  • Graphical leveling device provides warning when balance is not level
  • MinWeigh protection makes sure sample meets the minimum weight requirement for accuracy
  • TestManager embedded software keeps track of Standard Operating Procedures (SOPs) and alerts you when a test is due
  • Feedthrough for antitheft device and password protection for eight users enhance security
  • Built-in weighing modes include: percent weighing, dynamic weighing, statistics, piece counting, formulation and density determination, and factor calculation
  • StaticDetect™ new embedded technology automatically detects static charges
  • SmartSens infrared sensors integrated into display terminal allow for hands-free operation
  • Color touch screen functions in 11 languages
The Excellence XPE balances have one built-in RS-232 interface, with an additional slot available to install another interface module. Available modules include RS-232, USB, Ethernet, PS/2 (for connecting keyboards and barcode readers), LocalCAN, and MiniMettler (for connecting with older systems).
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Calibration Type Capacity (g) Readability (mg)
Availability Pricing
Mettler Toledo XPE26 Excellenceplus XPE Micro Balance, 22g x 0.001mg RK-11337-41 Mfr # XPE26
COMPARE
Internal 22 0.01
Call For Price
Mettler Toledo XPE26DR Excellenceplus XPE Micro Balance, 5.1/22g x 0.002/0.01mg RK-11337-42 Mfr # XPE26DR
COMPARE
Internal 22 0.002
Call For Price
Mettler Toledo XPE56 Excellenceplus XPE Micro Balance, 52g x 0.001mg RK-11337-43 Mfr # XPE56
COMPARE
Internal 52 0.01
Call For Price
Mettler Toledo XPE56DR Excellenceplus XPE Dr Micro Balance, 11/52g x 0.002/0.01mg RK-11337-44 Mfr # XPE56DR
COMPARE
Internal 52 0.002