Mettler Toledo Classic Light PL Portable Toploading Balances
Superior performance with easy operation
  • Mettler Toledo designed load cell guaranteeing accurate results
  • Large illuminated display for easy legibility under any light conditions
  • Easily understood user interface with icon driven functions
  • Molded cover allowing for stacking of up to 5 balances for quick and easy storage
  • Weighing result can be displayed in two units on measure at the same time
  • Unique environment setting allows the user to match the balance to the ambient conditions
  • Designed to be powered by battery or AC/DC adaptor for convenient portable operation
  • Vibration-resistant design ideal wieghing on temporary work surfaces or in the field
  • Transmits weighing results to a printer or a PC with the built-in RS232 interface
  • Feature low-battery, overload and stability indicators
  • Aviable weighing functions include; weighing, parts counting, percent weighing, check weighing, dynamic weighing, density along with several other modes
$2,114.31 - $2,519.50CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Mettler Toledo PL602E Classic Light Pl-E Portable Toploading Balance, 620G x 0.01g RK-11335-41 Mfr # PL602E
COMPARE
External 620 0.01
$2,114.31CAD / Each
Mettler Toledo PL1502E Classic Light Pl-E Portable Toploading Balance, 1520G x 0.01g RK-11335-42 Mfr # PL1502E
COMPARE
External 1520 0.01
$2,519.50CAD / Each
Mettler Toledo PL6001E Classic Light Pl-E Portable Toploading Balance, 6200G x 0.1g RK-11335-43 Mfr # PL6001E
COMPARE
External 6200 0.1
$2,114.31CAD / Each