This product is now discontinued, View Related Products

META Fluid-Resistant Women's Lab Coats

Liquid quickly beads and rolls off fabric, helping to keep lab coats dry and stain-free!
Lab coats are constructed of 65% polyester/35% cotton fabric and treated with NANO-TEX™ protection at a molecular level. Molecular structures, known as nano-whiskers, are attached to the tiny fabric fibers—permanently altering the performance of the fabric. This allows the fabric to repel liquids and stains without altering the look, feel, or comfort of the lab coat. Ideal for biotech, pharmaceutical, and general laboratory use.
Coats feature a five-button front and one interior (lower right) pocket to maximize functionality. One-piece back includes a two-button swing belt for added style and a better fit. Coats also feature three exterior pockets and one interior (lower right) pocket to maximize functionality. White collar.
$86.93CAD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Color Length (in) Size
Availability Pricing
Meta 17010-011-S Labcoat, Fluid Resistant, Ladies, S RK-86468-01 Mfr # 17010-011-S
COMPARE
White 40 Small
$86.93CAD / Each
Meta 17010-011-M Labcoat, Fluid Resistant, Ladies, M RK-86468-03 Mfr # 17010-011-M
COMPARE
White 40 Medium
$86.93CAD / Each
Meta 17010-011-L Labcoat, Fluid Resistant, Ladies, L RK-86468-05 Mfr # 17010-011-L
COMPARE
White 40 Large
$86.93CAD / Each
Meta 17010-011- Labcoat, Fluid Resistant, Ladies, XL RK-86468-07 Mfr # 17010-011-XL
COMPARE
White 40 X-Large
$86.93CAD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat