This product is now discontinued, View Related Products
Meriam MFC5150 HART-Compliant Communicator Series
Intuitive HART communicator makes commissioning any device fast and easy
  • HART-compliant modem communicates with any registered or unregistered device without translations
  • Touch screen menus operate without a stylus and are compatible with gloved hands
  • Full QWERTY keyboard and dedicated numeric keypad for easy device commissioning
  • Hot-key menu provides faster parameter modifications
  • 4 GB microSD memory card stores parameter configuration settings to facilitate duplication or restoration
  • Diagonal 4.3" display with white LED backlight for low-light conditions
  • Rechargeable Li-ion battery provides 10 hours continuous operation or up to 200 hours on standby
The MFC5150 HART communicator can be updated with new device information and firmware updates via the docking station's PC connection. It supports device specific commands as well as Universal and Common Practice communication with HART5, 6 and 7 devices.
Other convenient features include eight selectable languages, both Auto Dim and Auto Standby modes to conserve battery, adjustable backlight intensity, and the complete ISO Latin-1 character set via touch screen.
2 PRODUCT OPTIONS
Compareitem specifications
Max Pressure Pressure Accuracy Pressure Resolution
Availability Pricing
Meriam MFC5150 HART-Compliant Communicator RK-16108-14 Mfr # MFC5150
COMPARE
$8,250.98CAD / Each
Meriam MFC5150X HART-Compliant Communicator, intrinsically safe RK-16108-15 Mfr # MFC5150X
COMPARE
$8,825.61CAD / Each