This product is now discontinued, View Related Products
Megger MIT 400 Series Insulation Testers
For industrial, telecommunications, and special applications
  • Insulation testing to 1000 V and to 200 G ohms
  • Continuity testing at 200 mA down to 0.01 ohms
  • Analog and dual digital display
  • Insulation & continuity limit alarms PI/DAR timer function
  • Voltage measurements to 600 V True RMS
  • Test data storage and transfer by Compact Flash or Bluetooth® downloading
  • AC/DC voltage measurement
The Megger MIT400 Series of industrial insulation and continuity testers combine the latest range of measurement techniques with instrument design to provide a state-of-the-art tester that is compact and comfortable to hold.
$1,007.80 - $1,657.56CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Max Resistance (G ohms) Description
Availability Pricing
Megger MIT400-EN CAT IV 600V Basic Insulation Tester, 250-1000V RK-20022-38 Mfr # MIT400-EN
COMPARE
20 CAT IV 600V Basic Insulation Tester, 250-1000V
$1,007.80CAD / Each
Megger MIT410-EN CAT IV 600V Basic Insulation Tester, 50-1000V RK-20022-39 Mfr # MIT410-EN
COMPARE
100 CAT IV 600V Basic Insulation Tester, 50-1000V
$1,082.94CAD / Each
Megger MIT420-EN CAT IV 600V Advanced Insulation Tester, Data Storage RK-20022-40 Mfr # MIT420-EN
COMPARE
200 CAT IV 600V Advanced Insulation Tester, Data Storage
$1,321.63CAD / Each
Megger MIT430-EN CAT IV 600V Advanced Insulation Tester, Bluetooth RK-20022-41 Mfr # MIT430-EN
COMPARE
200 CAT IV 600V Advanced Insulation Tester, Bluetooth
$1,384.99CAD / Each
Megger MIT40X-EN Special 10-100V Selectable Insulation Tester RK-20022-42 Mfr # MIT40X-EN
COMPARE
20 Special 10-100V Selectable Insulation Tester
$1,298.06CAD / Each
Megger MIT480-EN Insulation Tester Cativ 600V Telecoms RK-20022-43 Mfr # MIT480-EN
COMPARE
100 Insulation Tester Cativ 600V Telecoms
$1,097.68CAD / Each
Megger MIT481-EN Insulation Tester Cativ 600V Telecom 5 and Storage RK-20022-44 Mfr # 1006-752
COMPARE
200 Insulation Tester Cativ 600V Telecom 5 and Storage
$1,561.79CAD / Each
Megger MIT485-EN CAT IV 600V Insulation Tester, Telecom 5 and Bluetooth RK-20022-45 Mfr # MIT485-EN
COMPARE
200 CAT IV 600V Insulation Tester, Telecom 5 and Bluetooth
$1,657.56CAD / Each
Megger MIT410-TC Insulation Tester, CATIII 600 V RK-20033-74 Mfr # MIT410-TC2
COMPARE
100 Insulation Tester, CATIII 600 V
$1,112.41CAD / Each
Megger MIT430-TC Insulation Tester, CATIV 600 V RK-20033-76 Mfr # 1000-574
COMPARE
200 Insulation Tester, CATIV 600 V
$1,407.09CAD / Each
Showing 1- 10 of 10