This product is now discontinued, View Related Products
Megger 5 kV and 10 kV Megohmmeters
Provides a continuously variable test voltage for highly sensitive insulation-resistance measurements of all types of electrical equipment and power cable
  • Provides 2% kV and M Ohm accuracy guard terminal to eliminate surface leakage current from the measurement Zero-start, high-voltage interlock Shielded, high-voltage output cable permanently connected to the instrument Circuit-breaker protection Redundant grounding connections Easy-to-follow, color-coded meter scales Analog metering for monitoring of variable measurements Rugged field construction
The units are suitable for dielectric absorption and step-voltage testing. Two models are available:The 5-kV Megohmmeter measures up to 100,000 MΩ; provides output from 250 volts to 5 kV, with direct reading up to 50,000 MΩ at 2.5 kV.The 15-kV Megohmmeter provides direct reading measure-ments up to 100,000 MΩ at 5 kV and up to 200,000 MΩ at 10 kV, with a multiplying factor for all other voltages up to 15 kV.The 15-kV model also features an internal voltage stabilizer that may be activated for improved measurement under extreme field conditions. Separate on/off controls are provided for power and high voltage, and a red light indicates when high voltage is on.
$9,775.94 - $12,862.69CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts)
Availability Pricing
Megger 210400 Megohmmeter Model , 5000 V DC, 100,000 M Ohms RK-20018-38 Mfr # 210400
COMPARE
5000
$9,775.94CAD / Each
Megger 210415 Megohmmeter Model , 15,000 V DC, 300,000 M Ohms RK-20018-39 Mfr # 210415
COMPARE
15000
$12,862.69CAD / Each