Masterflex® Ultrapure In-Line Flowmeters for Liquids
Precisely measure ultrapure water flow rates
These flowmeters use only plastic wetted parts, making them great for ultrapure water such as deionized water. End fittings are polyvinylchloride.
$482.68 - $539.41CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material Fitting Max Flow Rate (GPM) Min Flow Rate (GPM)
Availability Pricing
COMPARE
PVC 2 0.2
$482.68CAD / Each
COMPARE
PVC 5 0.5
$539.41CAD / Each