Masterflex® Transfer Tubing, Tygon® S3™ B-44-3, Non-DEHP Food and Beverage

Maintain sample purity
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Formulation: Tygon S3 B-44-3
Applications: Food and beverage processing and dispensing
Characteristics: Bio-based, phthalate-free flexible. Nonwetting, odorless, tasteless, and nontoxic. Clear
Certifications: FDA compliant, NSF-listed (Standard 51), and meets 3A Sanitary standards (20-27) for food and beverage handling
Temperature range: — 49 to 165°F ( — 45 to 74°C)
Sterilize: Ethylene oxide or autoclave for 30 min at 250°F (121°C), 15 psi (1 bar)
$67.04 - $769.11CAD / Pkg of 1

10 variations of this product are available.

See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
100 1/16 3/16
$67.04CAD / Pkg of 1
COMPARE
60 1/8 1/4
$92.82CAD / Pkg of 1
COMPARE
43 3/16 5/16
$144.39CAD / Pkg of 1
COMPARE
34 1/4 3/8
$163.55CAD / Pkg of 1
COMPARE
28 5/16 7/16
$193.01CAD / Pkg of 1
COMPARE
25 3/8 1/2
$221.01CAD / Pkg of 1
COMPARE
34 1/2 3/4
$468.54CAD / Pkg of 1
COMPARE
29 5/8 7/8
$565.78CAD / Pkg of 1
COMPARE
25 3/4 1
$611.46CAD / Pkg of 1
COMPARE
20 1 1 1/4
$769.11CAD / Pkg of 1
Showing 1- 10 of 10

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat