Masterflex® Transfer Tubing, Tygon® ND-100-65, Non-DEHP

Drains completely
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Formulation: ND-100-65
Applications: Pharmaceutical and biotech research applications.
Characteristics:Non-DEHP, phthalate free. Non-wetted surface permits complete drainage. Bondable, weldable. Clear.
Certification: USP Class VI, REACH, ISO 10993
Temperature range: -44 to 165°F (-42 to 74°C)
Sterilize: Autoclave, ethylene oxide, or irradiation.
$20.92 - $658.61CAD / Pkg of 1

25 variations of this product are available.

See All Product Options Below
25 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
100 1/32 3/32
$20.92CAD / Pkg of 1
COMPARE
55 1/16 1/8
$31.68CAD / Pkg of 1
COMPARE
40 3/32 5/32
$40.52CAD / Pkg of 1
COMPARE
55 1/8 1/4
$102.40CAD / Pkg of 1
COMPARE
45 5/32 9/32
$125.24CAD / Pkg of 1
COMPARE
20 3/16 1/4
$69.62CAD / Pkg of 1
COMPARE
55 3/16 3/8
$218.06CAD / Pkg of 1
COMPARE
70 3/16 7/16
$299.10CAD / Pkg of 1
COMPARE
18 1/4 5/16
$83.98CAD / Pkg of 1
COMPARE
30 1/4 3/8
$181.23CAD / Pkg of 1
COMPARE
45 1/4 7/16
$278.47CAD / Pkg of 1
COMPARE
55 1/4 1/2
$316.78CAD / Pkg of 1
COMPARE
25 5/16 7/16
$209.22CAD / Pkg of 1
COMPARE
20 3/8 1/2
$249.00CAD / Pkg of 1
COMPARE
30 3/8 9/16
$299.10CAD / Pkg of 1
COMPARE
40 3/8 5/8
$418.44CAD / Pkg of 1
COMPARE
18 1/2 5/8
$285.84CAD / Pkg of 1
COMPARE
25 1/2 11/16
$380.13CAD / Pkg of 1
COMPARE
30 1/2 3/4
$480.33CAD / Pkg of 1
COMPARE
25 5/8 7/8
$607.04CAD / Pkg of 1
Showing 1 - 20 of 25

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat