Masterflex® Transfer Tubing, Tygon® E-3603, Non-DEHP
Pliable formulation for sharp radius bending
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. Temperature range is -51 to 165°F (-46 to 73°C). Can be sterilized with ethylene oxide or autoclave.
$140.71 - $3,232.62CAD / Pkg of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
36 1/8 1/4
$966.54CAD / Pkg of 1
COMPARE
20 1/4 3/8
$1,508.75CAD / Pkg of 1
COMPARE
36 1/4 1/2
$3,232.62CAD / Pkg of 1
COMPARE
30 5/16 9/16
$2,009.70CAD / Pkg of 1
COMPARE
69 1/32 3/32
$140.71CAD / Pkg of 1
COMPARE
36 1/16 1/8
$262.26CAD / Pkg of 1