Masterflex® Transfer Tubing, Peroxide-Cured Reinforced Silicone
Low compression formulation keeps its shape and cleanliness
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Applications: Demanding high-pressure pharmaceutical, biotechnology, aerospace, and food and beverage applications.
Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, low tackiness, translucent.
Certifications: Elastomer is FDA compliant and meets USP Class VI. Tubing is manufactured according to GMP.
Temperature range: -60 to 400°F (-51 to 204°C)
Sterilize: Autoclave or gamma irradiation.
$928.24 - $2,817.12CAD / Pkg of 1
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
175 1/8 3/8
$928.24CAD / Pkg of 1
COMPARE
155 1/4 9/16
$1,193.45CAD / Pkg of 1
COMPARE
145 5/16 19/32
$1,440.98CAD / Pkg of 1
COMPARE
138 3/8 11/16
$1,491.07CAD / Pkg of 1
COMPARE
125 1/2 13/16
$2,121.68CAD / Pkg of 1
COMPARE
57 3/4 1
$2,817.12CAD / Pkg of 1
COMPARE
119 7/8 1 1/4
$2,787.65CAD / Pkg of 1
COMPARE
113 1 1
$1,258.28CAD / Pkg of 1
Showing 1- 8 of 8