Masterflex® Transfer Tubing, Perfluoroalkoxy (PFA)
Stands up to highly corrosive materials
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. For the transfer of high-temperature or highly corrosive materials. Also good for high-purity work.
Characteristics: Chemically inert, nontoxic, and virtually nonporous. Excellent crack- and stress-resistance. Low coefficient of friction. Translucent.
Resin certification: Resin is FDA-compliant (21 CFR 177.1550); meets USP Class VI requirements. Tubing is manufactured according to GMP.
Temperature range: -320 to 500°F (-195 to 260°C)
Sterilize: Autoclave, ethylene oxide, or use dry heat.
$64.83 - $1,001.91CAD / Pkg of 1
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
600 1/32 1/16
$64.83CAD / Pkg of 1
COMPARE
471 3/32 5/32
$109.03CAD / Pkg of 1
COMPARE
385 1/8 3/16
$128.92CAD / Pkg of 1
COMPARE
280 3/16 1/4
$159.13CAD / Pkg of 1
COMPARE
220 1/4 5/16
$206.27CAD / Pkg of 1
COMPARE
180 5/16 3/8
$278.47CAD / Pkg of 1
COMPARE
117 1/32 9/16
$386.03CAD / Pkg of 1
COMPARE
471 3/16 5/16
$366.87CAD / Pkg of 1
COMPARE
229 19/64 3/8
$384.55CAD / Pkg of 1
COMPARE
324 5/16 7/16
$474.43CAD / Pkg of 1
COMPARE
220 9/16 1/2
$403.71CAD / Pkg of 1
COMPARE
220 1/2 5/8
$702.81CAD / Pkg of 1
COMPARE
87 11/16 3/4
$545.15CAD / Pkg of 1
COMPARE
153 3/4 7/8
$1,001.91CAD / Pkg of 1
COMPARE
592 1/16 1/8
$70.72CAD / Pkg of 1
COMPARE
421 5/32 1/4
$165.02CAD / Pkg of 1
COMPARE
300 7/32 5/16
$390.45CAD / Pkg of 1
COMPARE
388 1/4 3/8
$363.93CAD / Pkg of 1
COMPARE
283 3/8 1/2
$437.60CAD / Pkg of 1
COMPARE
150 5/8 3/4
$651.24CAD / Pkg of 1
Showing 1 - 20 of 24