Masterflex® Transfer Tubing, Fluorinated Etyhlene Propylene (FEP)
Does not deteriorate with age
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Excellent for the transfer of extreme temperature or highly corrosive materials. Also good for high-purity work.
Characteristics: Chemically inert, nontoxic, heat resistant, with a low coefficient of friction. Doesn’t deteriorate with age. Tough, but bends even at -454°F (-270°C). Translucent.
Certifications: Resin is FDA-compliant (21 CFR 177.1550). Tubing is manufactured in ISO 9001-2008 approved facility.
Temperature range: -454 to 400°F (-270 to 205°C)
Sterilize: Autoclave, ethylene oxide, or use dry heat.
$29.17 - $580.52CAD / Pkg of 1
23 variations of this product are available.
See All Product Options Below
23 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
110 3/8 7/16
$109.77CAD / Pkg of 1
COMPARE
88 1/2 9/16
$179.75CAD / Pkg of 1
COMPARE
500 1/32 1/16
$29.17CAD / Pkg of 1
COMPARE
444 1/16 1/8
$34.26CAD / Pkg of 1
COMPARE
150 1/4 5/16
$85.46CAD / Pkg of 1
COMPARE
140 5/16 3/8
$105.35CAD / Pkg of 1
COMPARE
102 7/16 1/2
$163.55CAD / Pkg of 1
COMPARE
71 5/8 11/16
$268.16CAD / Pkg of 1
COMPARE
65 11/16 3/4
$291.73CAD / Pkg of 1
COMPARE
172 19/64 3/8
$265.21CAD / Pkg of 1
COMPARE
329 5/32 1/4
$146.60CAD / Pkg of 1
COMPARE
450 1/8 1/4
$166.49CAD / Pkg of 1
COMPARE
353 3/16 5/16
$201.85CAD / Pkg of 1
COMPARE
288 1/4 3/8
$254.90CAD / Pkg of 1
COMPARE
243 5/16 7/16
$330.04CAD / Pkg of 1
COMPARE
210 3/8 1/2
$378.66CAD / Pkg of 1
COMPARE
185 7/16 9/16
$368.35CAD / Pkg of 1
COMPARE
165 1/2 5/8
$447.91CAD / Pkg of 1
COMPARE
135 5/8 3/4
$565.78CAD / Pkg of 1
COMPARE
115 3/4 7/8
$580.52CAD / Pkg of 1
Showing 1 - 20 of 23