Masterflex® Microscope Slides with Printed Sections
Optimize sample testing and viewing
  • Screen-printed section design keeps samples contained
  • Hydrophobic epoxy ink withstands frequent washing and resists chemicals
  • Contrasting ink makes orienting the microscope field of view over a sample easy
Printed microscope slides are available with a variety of section shapes, sizes, counts, colors, and labels. The PTFE-based epoxy ink creates a hydrophobic barrier, holding samples in place for easy viewing under a microscope, and keeping reactions isolated from each other in multi-section slides.
$94.30 - $222.48CAD / Pkg of 144
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Slide Type Description
Availability Pricing
COMPARE
Printed without Mask Printed Microscope Slide, ten rectangular sections; 144/pk
$97.98CAD / Pkg of 144
COMPARE
Printed without Mask Printed Microscope Slide, one rectangular section; 144/pk
$94.30CAD / Pkg of 144
COMPARE
Printed with Mask Printed Microscope Slide, one rectangular section with mask; 144/pk
$94.30CAD / Pkg of 144
COMPARE
Printed without Mask Printed Microscope Slide, one circle, 5 mm; 144/pk
$94.30CAD / Pkg of 144
COMPARE
Printed without Mask Printed Microscope Slide, two circles, 8 mm, positive/negative; 144/pk
$94.30CAD / Pkg of 144
COMPARE
Printed with Mask Printed Microscope Slide, two circles, 7 mm, positive/negative; 144/pk
$94.30CAD / Pkg of 144
COMPARE
Printed with Mask Printed Microscope Slide, gridded with mask; 144/pk
$222.48CAD / Pkg of 144
Showing 1- 7 of 7