Masterflex® Mass Flowmeter Controllers for Gases
Integrated valve allows precise flow control
  • Accepts a 0 to 5 VDC input signal for remote valve control
  • Calibrated for air and nitrogen
These low-cost mass flowmeter controllers use a thermal gas flow sensing technique that results in highly accurate readings and repeatability. The controllers accept a 0 to 5 VDC input signal for remote valve control. Controllers with LCD display feature a potentiometer for manual valve control. The 3 1/2"-digit display tilts up to 90° for easy viewing. Front panel dip switch lets you select one of the factory programmed gases: air or nitrogen.
All controllers require a combined power supply/input/output cable. This cable combines three functions into one unit: power source, plus input or output of 0 to 5 VDC signals. Use the input signal for remote valve control; use the output signal for datalogging or controlling other instruments. Order the power supply/input/output cable separately.
Wetted materials are anodized aluminum, 316 stainless steel, brass, Viton®, and acetal. Maximum pressure drop across the unit is 15 psi at maximum flow. Differential pressure should not exceed 45 psi.
These precalibrated flowmeters operate at inlet pressures between 5 and 40 psi and at gas temperatures between 64 and 77°F (18 to 25°C) while maintaining the stated ±1.5% full scale accuracy and linearity. When operating beyond 5 to 40 psi, add ±0.02%/psi full scale; if operating beyond 64 to 77°F (18 to 25°C), add ±0.15%/°C full scale.
$1,464.55 - $1,986.87CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Display Type Gas Type Max Flow Rate (LPM)
Availability Pricing
COMPARE
None Air, Nitrogen 0.05
$1,464.55CAD / Each
COMPARE
None Air, Nitrogen 0.1
$1,631.78CAD / Each
COMPARE
None Air, Nitrogen 0.5
$1,580.21CAD / Each
COMPARE
None Air, Nitrogen 1
$1,558.11CAD / Each
COMPARE
None Air, Nitrogen 5
$1,614.10CAD / Each
COMPARE
3.5-digit LCD Air, Nitrogen 0.05
$1,801.96CAD / Each
COMPARE
3.5-digit LCD Air, Nitrogen 1
$1,986.87CAD / Each
COMPARE
3.5-digit LCD Air, Nitrogen 5
$1,986.87CAD / Each
COMPARE
3.5-digit LCD Air, Nitrogen 10
$1,975.82CAD / Each
Showing 1- 9 of 9