Masterflex® Low Pressure-Drop Liquid Flow Switches
Minimize flow constriction and achieve low pressure drop
  • Chemically resistant — wetted materials Ryton® PPS, Viton®, 316 Stainless Steel, and polypropylene
  • Effortlessly install with 1/2" NPT(M) connections
  • Durable switch with 212°F (100°C) max temperature and 200 psi at 70°F max pressure
These SPST switches feature a unique, dual-diameter, internal bore and piston configuration to minimize flow constriction and provide low pressure drop. Use a 150 µm filter to protect your switch. Switches include 1/4" male quick connect terminals.
$176.81CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Normally closed switch for liquids; 0.5 GPM
$176.81CAD / Each
COMPARE
Normally closed switch for liquids; 1.0 GPM
$176.81CAD / Each
COMPARE
Normally closed switch for liquids; 2.0 GPM
$176.81CAD / Each
COMPARE
Normally closed switch for liquids; 5.0 GPM
$176.81CAD / Each