Masterflex® Liquid Flow Switches for Threaded Plastic Piping
Connects quickly to threaded plastic piping
  • Chemically resistant — wetted materials stainless steel and polypropylene
  • Effortlessly install with 1/2" NPT(M) connections
  • NSF and FDA approved
These normally open switches have 3/4" NPT(F) ports for a quick connection to threaded plastic piping. Polypropylene and stainless steel wetted parts offer excellent chemical resistance. The materials are also NSF approved and FDA compliant; approved for potable water treatment applications such as chlorinators, purifiers, and heaters. Switches include 24" lead wires.
$145.87 - $203.33CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Min Flow Rate (GPM) Pipe Size
Availability Pricing
COMPARE
$145.87CAD / Each
COMPARE
$203.33CAD / Each
COMPARE
$203.33CAD / Each
COMPARE
$145.87CAD / Each
COMPARE
$145.87CAD / Each
COMPARE
$145.87CAD / Each