This product is now discontinued, View Related Products
Masterflex L/S Precision Solve-Flex® PTFE Pump Tubing
High-performance pumpable fluoropolymer (PTFE) tubing for challenging applications
  • Superb chemical resistance
  • Acid-resistant, solvent-resistant
  • Flexible tubing construction
  • Long, continuous lengths; allow for unobstructed fluid path
  • Meets USP Class VI criteria
Choose our Solve-Flex® PTFE tubing for handling the toughest chemical applications. Ideal for solvents and strong acids, this flexbile peristlatic tubing is chemically compatible with hydrofluoric acid, MEK and sulfurous acid.
Based on your application needs, choose Precision tubing or High-Performance Precision pump tubing. High-Performance Precision tubing has a thicker wall that improves pressure generation suction lift, and the ability to pump viscous fluids. Solve-Flex® tubing has a working temperature range of -20 to 100°C (-4 to 212°F).
Tubing can be sterilized by EtO.
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) L/S Masterflex tubing size Max Flow Rate (mL/min)
Availability Pricing
COMPARE
1/8 16 480
$1,006.33CAD / Pkg
COMPARE
1/4 17 1700
$1,050.53CAD / Pkg of 1
COMPARE
1/4 24 1800
$1,199.34CAD / Pkg of 1