Masterflex® L/S® Precision Pump Tubing, BioPharm Platinum-Cured Silicone
Ultra-smooth inner surface minimizes particle entrapment
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: Pharmaceutical, cosmetic, research, and auto-analysis applications
Characteristics: Ultra-smooth, minimizes bacterial growth, good for mild to medium concentration bases, salts, and alcohols; odorless, tasteless, nontoxic, transparent
$143.66 - $335.93CAD / Pkg of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) L/S Masterflex tubing size Max Flow Rate (mL/min)
Availability Pricing
COMPARE
1/16 13 36
$143.66CAD / Pkg of 1
COMPARE
1/16 14 130
$165.02CAD / Pkg of 1
COMPARE
1/8 16 480
$191.54CAD / Pkg of 1
COMPARE
1/4 17 1700
$253.42CAD / Pkg of 1
COMPARE
3/8 18 2300
$335.93CAD / Pkg of 1
COMPARE
3/16 25 1000
$231.32CAD / Pkg of 1