Masterflex® L/S® High-Performance Precision Pump Tubing, Norprene® Food A-60-F
Similar to Norprene® but with FDA approval
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: Ideal for the food, dairy, and beverage industries
Characteristics: Heat, ozone, and UV light resistant, nonaging, nonoxidizing, superior acid and alkali resistance, opaque beige
$212.17 - $324.15CAD / Pkg of 1
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) L/S Masterflex tubing size Max Flow Rate (mL/min)
Availability Pricing
COMPARE
3/16 15 1100
$212.17CAD / Pkg of 1
COMPARE
1/4 24 1800
$274.05CAD / Pkg of 1
COMPARE
3/8 35 2600
$300.57CAD / Pkg of 1
COMPARE
3/8 36 3400
$324.15CAD / Pkg of 1