Masterflex® L/S® Easy-Load® II Pump Heads with 2-Channels for Precision Tubing
No need to stack heads—get two channels in one head
  • New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
  • Four-roller rotor improves pressure performance and mechanical stability while reducing pulsation
  • Compact design for efficient use of space
  • Suitable for use with L/S® Precision pump tubing sizes
The two-channel pump head lets you pump two synchronous flow channels, proper retention requires the use of the same size tubing in each channel. These pump heads accept the six L/S® precision tubing sizes L/S® 13, L/S® 14, L/S® 16, and L/S® 25. Achieve flow rates of 0.06 to 1000 mL/min—flow rate depends on drive rpm and tubing size.
$592.30 - $701.33CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate Per Channel (mL/min) Rotor Material Tubing sizes accepted
Availability Pricing
COMPARE
1000 Cold-rolled Steel L/S® 13, L/S 14, L/S 16, L/S 25
$592.30CAD / Each
COMPARE
1000 Stainless Steel L/S® 13, L/S 14, L/S 16, L/S 25
$701.33CAD / Each