Masterflex® Impact-Resistant Direct-Reading Variable-Area Flowmeters for Gas and Water, Polycarbonate Housing
Precision-adjusting valve for accurate, leak-free flow control
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Constructed with durable polycarbonate frame for rugged applications
  • Designed with graduations in English and metric for additional flexibility
Molded of high-impact, resistant polycarbonate, these direct-reading flowmeters are supplied with scales in both English and metric units for liquid and air, all with 10:1 turndown ratios. These flowmeters has been designed to maintain maximum pressures to 100 psig and temperatures to 150°F. Flowmeters are ideal for OEM applications.
$104.76 - $139.38CAD / Each
26 variations of this product are available.
See All Product Options Below
26 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fluid Type Max Flow Rate (LPM) Valve
Availability Pricing
COMPARE
Water 0.157 No
$117.28CAD / Each
COMPARE
Water 0.631 No
$121.92CAD / Each
COMPARE
Water 2.524 No
$104.76CAD / Each
COMPARE
Water 0.11 No
$104.76CAD / Each
COMPARE
Water 0.3 No
$112.71CAD / Each
COMPARE
Air 1.18 No
$117.28CAD / Each
COMPARE
Air 5.191 No
$112.71CAD / Each
COMPARE
Air 28.317 No
$104.76CAD / Each
COMPARE
Air 1.2 No
$104.76CAD / Each
COMPARE
Air 5 No
$112.71CAD / Each
COMPARE
Air 10 No
$104.76CAD / Each
COMPARE
Air 50 No
$104.76CAD / Each
COMPARE
Air 100 No
$112.71CAD / Each
COMPARE
Water 0.157 Yes
$130.76CAD / Each
COMPARE
Water 0.631 Yes
$134.74CAD / Each
COMPARE
Water 0.3 Yes
$134.74CAD / Each
COMPARE
Air 1.2 Yes
$134.74CAD / Each
COMPARE
Air 5 Yes
$134.74CAD / Each
COMPARE
Air 10 Yes
$134.74CAD / Each
COMPARE
Air 100 Yes
$134.74CAD / Each
Showing 1 - 20 of 26