Masterflex® Drum Pump Air Motors
Safety transfer a variety of liquids from drums, tanks, and small storage vessels
  • Maintenance is reduced with no mechanical seals
  • Run dry capability
Combine the dump pump air motors with a flowmeter (sold separately) to display current and total volume dispensed, Each motor includes a muffler. A filter, regulator, and lubricator are recommended.
$1,016.64 - $1,290.69CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Max Motor RPM (rpm) Description
Availability Pricing
COMPARE
20 7500 Drum Pump Air Motor, 22 cfm at 92 psi
$1,016.64CAD / Each
COMPARE
30 8000 Drum Pump Air Motor, 28 cfm at 90 psi
$1,290.69CAD / Each