Masterflex® Direct-Reading Variable-Area Flowmeters for Gases and Water, Acrylic
Designed for labs and plant processes where multiple gases are used
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
Versatile acrylic flowmeters are designed specifically for air, oxygen, nitrogen, carbon dioxide, argon, helium, or water. Flowmeters are offered with or without built-in low hysteresis needle valves. Panel mount flowmeters without valves or use in partial or full in-line mounted configurations. Flowmeters feature a stable, easy-to-read float for quick measurement readings.
$75.14 - $112.20CAD / Each
196 variations of this product are available.
See All Product Options Below
196 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Fitting Max Flow Rate (LPM) Valve
Availability Pricing
COMPARE
316 SS 1.4 Yes
$112.20CAD / Each
COMPARE
316 SS 8.5 Yes
$112.20CAD / Each
COMPARE
316 SS 2.5 No
$85.68CAD / Each
COMPARE
316 SS 3.5 No
$85.68CAD / Each
COMPARE
316 SS 15 No
$85.68CAD / Each
COMPARE
316 SS 20 No
$85.68CAD / Each
COMPARE
316 SS 45 No
$85.68CAD / Each
COMPARE
Brass 1.2 Yes
$90.10CAD / Each
COMPARE
Brass 20 Yes
$90.10CAD / Each
COMPARE
Brass 7 No
$78.31CAD / Each
COMPARE
Brass 1.1 No
$78.31CAD / Each
COMPARE
316 SS 5.5 Yes
$112.20CAD / Each
COMPARE
316 SS 8 Yes
$112.20CAD / Each
COMPARE
Brass 0.02 No
$78.31CAD / Each
COMPARE
Brass 0.07 No
$78.31CAD / Each
COMPARE
Brass 0.1 No
$78.31CAD / Each
COMPARE
316 SS 0.02 No
$85.68CAD / Each
COMPARE
316 SS 0.7 No
$85.68CAD / Each
COMPARE
Brass 0.07 Yes
$90.10CAD / Each
COMPARE
316 SS 1.4 Yes
$112.20CAD / Each
Showing 1 - 20 of 196