Masterflex® Direct- Reading Variable-Area Flowmeter Kits with Interchangeable Gas and Water Scales
Seven interchangeable scales for routine gases and water offer flexibility
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Designed with stable, easy-to-read float for quick measurement readings
  • Interchangeable scales offer English and metric scales
  • Use with multiple routine gases for laboratory and industrial applications
  • Easy disassembly and assembly for cleaning
These easy-to-read acrylic flowmeter kits come with seven interchangeable back plates for routine gases air, argon, nitrogen, helium, carbon dioxide, oxygen, and water. Meters are offered with or without built-in low hysteresis needle valves. Meters without valves can be panel mounted or used in partial or full in-line mounted configurations.
$107.85 - $141.89CAD / Each
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
Compareitem specifications
Material Fitting Max Flow Rate (mL/min) Description
Availability Pricing
COMPARE
Brass 1200 Flowmeter Kit, Brass Fittings; 1.2 LPM Air
$107.85CAD / Each
COMPARE
Brass 1200 Flowmeter Kit, Brass Fittings; 1.2 LPM Air With Valve
$119.64CAD / Each
COMPARE
SS 1200 Acrylic Flowmeter Kit, SS, 0.02 LPM Air / .3 GPH Water
$115.22CAD / Each
COMPARE
SS 1200 Flowmeter Kit, SS Fittings; 1.2 LPM Air With Valve
$141.89CAD / Each
COMPARE
Brass 2500 Flowmeter Kit, Brass Fittings; 2.5 LPM Air
$107.85CAD / Each
COMPARE
Brass 2500 Flowmeter Kit, Brass Fittings; 2.5 LPM Air With Valve
$119.64CAD / Each
COMPARE
SS 2500 Acrylic Flowmeter Kit, SS, 2.5 LPM Air / 70 GPH Water
$115.22CAD / Each
COMPARE
Brass 3500 Flowmeter Kit, Brass Fittings; 3.5 LPM Air
$107.85CAD / Each
COMPARE
Brass 3500 Flowmeter Kit, Brass Fittings; 3.5 LPM Air With Valve
$119.64CAD / Each
COMPARE
SS 3500 Acrylic Flowmeter Kit, SS, 3.5 LPM Air / 100 GPH Water
$115.22CAD / Each
COMPARE
Brass 8500 Flowmeter Kit, Brass Fittings; 8.5 LPM Air
$107.85CAD / Each
COMPARE
Brass 8500 Flowmeter Kit, Brass Fittings; 8.5 LPM Air With Valve
$119.64CAD / Each
COMPARE
SS 8500 Acrylic Flowmeter Kit, SS, 8.5 LPM Air / 175 GPH Water
$114.04CAD / Each
COMPARE
SS 8500 Flowmeter Kit, SS Fittings; 8.5 LPM Air With Valve
$141.89CAD / Each
COMPARE
Brass 16000 Flowmeter Kit, Brass Fittings; 16 LPM Air
$107.85CAD / Each
COMPARE
SS 16000 Acrylic Flowmeter Kit, SS, 16 LPM Air / 450 GPH Water
$114.04CAD / Each
COMPARE
Brass 22000 Flowmeter Kit, Brass Fittings; 22 LPM Air
$107.85CAD / Each
COMPARE
Brass 22000 Flowmeter Kit, Brass Fittings; 22 LPM Air With Valve
$119.64CAD / Each
COMPARE
SS 22000 Acrylic Flowmeter Kit, SS, 22 LPM Air / 700 GPH Water
$115.22CAD / Each
COMPARE
SS 22000 Flowmeter Kit, SS Fittings; 22 LPM Air With Valve
$141.89CAD / Each
Showing 1 - 20 of 24