Masterflex® Digital Benchtop Gear Pump Systems
Order the complete pump drive and head system
  • Graphical LCD shows four operating modes
  • IP33 rating ensures splash and dust resistance
  • Systems deliver pulse-free flow in combination with Micropump® A-mount head
  • Remote control via DB25 female connector on drive
Calibrate, copy, and dispense features produce precise, pulseless batching and dispensing. View pump performance continually. Retrieve displacement and flow values by pump head size.
These precision systems are easily operated from the intuitive, multilanguage LCD front panel or optional foot switch 07523-92 for hands-free on/off control. The IP33-rated enclosure protects internal components from damage.
Service kits 07144-42 and 74012-52 include: gears, suction shoe, shaft seals, and springs
Service kit 74012-22 includes: gears, bushings, and seals
$4,178.53 - $4,744.32CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Differential Pressure (PSI) Wetted Parts Max Flow Rate (mL/min)
Availability Pricing
COMPARE
75 316 SS, PPS, PTFE 331.2
$4,558.67CAD / Each
COMPARE
75 316 SS, PPS, PTFE 1137.6
$4,442.27CAD / Each
COMPARE
75 316 SS, PPS, PTFE 2304
$4,467.32CAD / Each
COMPARE
75 316 SS, PPS, PTFE 3276
$4,420.17CAD / Each
COMPARE
60 316 SS, PPS, Viton 4212
$4,744.32CAD / Each
COMPARE
125 316 SS, PEEK, Viton 939.6
$4,529.20CAD / Each
COMPARE
75 316 SS, PPS, PPS 331.2
$4,178.53CAD / Each
COMPARE
20 316 SS, Graphite, PTFE 61
$4,437.85CAD / Each
Showing 1- 8 of 8