MAPA Butoflex 650 Cotton-Lined Butyl Gloves
Inner fabric lining provides exceptional comfort
Butyl gloves provide excellent chemical resistance in applications involving acids, ketones, esters, and amine derivatives. 100% seamless cotton liner provides extra comfort for long-term use. Scalloped cuffs can be rolled down to prevent drips from reaching your arms. Unique Z-pattern grip design for optimal grip.
$131.13 - $149.92CAD / Pair of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Size
Availability Pricing
MAPA Butoflex 650 Cotton-Lined Butyl Gloves, size 8, 1 pair RK-86357-41 Mfr # 65031-8
COMPARE
14 8
$131.13CAD / Pair of 1
MAPA Butoflex 650 Cotton-Lined Butyl Gloves, size 10, 1 pair RK-86357-43 Mfr # 6503110
COMPARE
14 10
$149.92CAD / Pair of 1