MagStir Genie Programmable Magnetic Stirrer
Microprocessor controls offer precision and reproducibility without internal heat build-up.
Low-profile magnetic stirrer uses microprocessor control for precise and reproducible operation without heat build-up from internal friction. Manual mode provides stir, stir reverse, and time functions. Profiles mode allows users to quickly program speed, ramp rate, stir direction, time (including pause), and power level in up to 9 distinct steps. Up to 4 stirring profiles can be saved and activated in automatic mode. Can be used with any regular flask/beaker or spinner flask. Speed range of 10-1800 RPM including reverse and interval stirring for applications ranging from gentle stirring for cell culture to aggressive mixing for viscous polymers. Three power levels available for various sample viscosities.
$949.60CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Scientific Industries Genie SI-0300 MagStir Compact Programmable Stirrer 120 VAC RK-86575-01 Mfr # SI-0300
COMPARE
120
$949.60CAD / Each