This product is now discontinued, View Related Products
M-SYSTEM DL8 Web-Based Recorders
Choose a recorder with the right combination of communication functions
Along with a router and web-enabled device, this system allows you to control, monitor and record any process by accessing a web browser. Data may also be sent by encrypted SMTP over SSL e-mail. E-mail functions as an event alert or a regularly scheduled report.
To complete your system, choose remote I/O modules and the R8 connector.
$884.03 - $1,178.71CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Temperature Accuracy (° F) Temperature Resolution (° F)
Availability Pricing
M-System DL8 Web-Enabled Recorder, Modbus/TCP ethernet, FTP client, e-mail RK-45004-11 Mfr # DL8-B-R
COMPARE
$884.03CAD / Each
M-System DL8 Web-Enabled Recorder, Modbus/TCP, FTP, e-mail, data logging RK-45004-12 Mfr # DL8-C-R
COMPARE
$1,031.37CAD / Each
M-System DL8 Web-Enabled Recorder, Modbus/TCP, FTP, email, data logging, I/O map RK-45004-13 Mfr # DL8-D-R
COMPARE
$1,178.71CAD / Each