This product is now discontinued, View Related Products

Lufkin Measuring Tapes

All tapes are manufactured to US government standards, are checked for accuracy under NIST standards
  • Durable lightweight Hi-Viz ABS case
  • Cushioned blade return extends product life
  • Bright yellow blade has clear and easy-to-read numerals
  • Convenient toggle lock holds blade securely in an position
  • Quikread® tape 97105-89 has graduations printed in fractions and decimal equivalents
$18.99 - $42.73CAD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (in) Width (in) Description
Availability Pricing
Lufkin 1" x 25' Hi-Viz Orange Power Tape RK-97105-86 Mfr # PHV1425N
COMPARE
300 1 1\" x 25' Hi-Viz Orange Power Tape
$42.73CAD / Each
Lufkin 3/4" x 16' Hi-Viz Orange Series 1000 Power Tape RK-97105-87 Mfr # HV1316
COMPARE
192 0.75 3/4\" x 16' Hi-Viz Orange Series 1000 Power Tape
$18.99CAD / Each
Lufkin 3/4" x 12' Hi-Viz Orange Series 1000 Power Tape RK-97105-88 Mfr # PHV1312N
COMPARE
144 0.75 3/4\" x 12' Hi-Viz Orange Series 1000 Power Tape
$24.68CAD / Each
Lufkin 1" x 25' Quickread Power Return Tape RK-97105-89 Mfr # PQR1425N
COMPARE
300 1 1\" x 25' Quickread Power Return Tape
$36.83CAD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat