MadgeTech Low Cost Temperature and Humidity Data Loggers
TransiTempII, TransiTempII-RH, and Cryo-Temp
  • Programmable alarms and warnings
  • Instantaneous out of range indication
  • IP65 splash proof rated
  • Delayed Start
These splash proof temperature data loggers featuring a highly precise semiconductor temperature sensor and an ABS plastic enclosure. This affordable, N.I.S.T. traceable device is ideal for monitoring and recording the temperature of all types of perishable products, including food items and pharmaceutical goods, both in storage and in transit.
The three available devices are equipped with three status LED indicators to alert you to the temperature status. Green signals safe conditions, yellow indicates warning limits have been exceeded, and red denotes alarm limits have been exceeded. The units can be programmed through software to start immediately, delayed start for up to 30 days, or be set to start using the magnetic switch.
$378.29CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Temperature Accuracy (° C) Temperature Resolution (° C)
Availability Pricing
MadgeTech Cryo-Temp Ultra-Low Temperature Data Logger RK-23000-27 Mfr # CRYO-TEMP
COMPARE
35 ±1.0 0.1
$378.29CAD / Each